Anti-ADCY7 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB419MU01
MSPP-PAB419MU1EA 342 USD
MSPP-PAB419MU1
Anti-ADCY7 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Adenylate Cyclase 7 (ADCY7), derived from recombinant ADCY7(Leu309~Glu491), is reactive with Human/Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Adenylate cyclase type 7 is a membrane-bound adenylate cyclase that catalyses the formation of cyclic AMP from ATP and is inhibitable by calcium. The product of this gene is a member of the adenylyl cyclase class-4/guanylyl cyclase enzyme family that is characterized by the presence of twelve membrane-spanning domains in its sequences.The deduced protein contains 12 membrane-spanning domains, a characteristic of the adenylyl cyclase enzyme family. RNase protection assays indicated that ADCY7 is the major form of adenylyl cyclase in human platelets. Northern blot analysis detected a 6.7-kb transcript that was abundant in liver. Of brain areas studied, the transcript was most abundant in the caudate and cerebellum and present at a slightly lower level in hippocampus.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: ADCY7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR