Anti-HB Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB409BO01
MSPP-PAB409BO1EA 294 USD
MSPP-PAB409BO1
Anti-HB Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Hemoglobin (HB), derived from native HB, is reactive with Human/Bovine.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Hemoglobin is the iron-containing oxygen-transport metalloprotein in the red blood cells of vertebrates, and the tissues of some invertebrates. In mammals, the protein makes up about 97% of the red blood cell’s dry content, and around 35% of the total content. Hemoglobin transports oxygen from the lungs or gills to the rest of the body where it releases the oxygen for cell use. It also has a variety of other roles of gas transport and effect-modulation which vary from species to species, and are quite diverse in some invertebrates.  Hemoglobin has an oxygen binding capacity of between 1.36 and 1.37 ml O2 per gram of hemoglobin, which increases the total blood oxygen capacity seventyfold. Hemoglobin is found in nonerythroid cells including the A9 dopaminergic neurons in the substantia nigra, macrophages, alveolar cells, and mesangial cells in the kidney.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: HB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR