Anti-LZM Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB193BO01
MSPP-PAB193BO1EA 400 USD
MSPP-PAB193BO1
Anti-LZM Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Lysozyme (LZM), derived from recombinant LZM(Gly18~Leu147), is reactive with Bovine.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Lysozyme, also known as muramidase or N-acetylmuramide glycanhydrolase, are a family of enzymes (EC 3.2.1.17) which damage bacterial cell walls by catalyzing hydrolysis of 1,4-beta-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in a peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrins.,Lysozyme is abundant in a number of secretions, such as tears, saliva, human milk and mucus. It is also present in cytoplasmic granules of the polymorphonuclear neutrophils (PMN). Large amounts of lysozyme can be found in egg white. C-type lysozymes are closely related to alpha-lactalbumin in sequence and structure making them part of the same family.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: LZM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR