Anti-CLU Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB180MU01
MSPP-PAB180MU1EA 224 USD
MSPP-PAB180MU1
Anti-CLU Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Clusterin (CLU), derived from recombinant CLU (Ser227~Glu448), is reactive with Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Clusterin (apolipoprotein J) is a 75 to 80 kDa disulfide-linked heterodimeric protein associated with the clearance of cellular debris and apoptosis. This protein has several synonyms: dimeric acidic glycoprotein (DAG protein), testosterone repressed prostate message-2 (TRPM-2), sulfated glycoprotein-2 (SGP-2) and complement lysis inhibitor (CLI). This protein was first identified in 1988 in seminal fluid. In humans the gene is encoded on chromosome 8 (8p21) and is highly conserved between species (70-80% homology). It is expressed in most mammalian tissues and can be found in blood plasma, milk, urine, cerebrospinal fluid and semen. A number of proteins have been found to affect its expression including Egr-1, members from the AP-1 complex, HSF1/2, Cdx1/2 and B-Myb.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: CLU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR