Anti-MT1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB119HU01
MSPP-PAB119HU1EA 332 USD
MSPP-PAB119HU1
Anti-MT1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Metallothionein 1 (MT1), derived from recombinant MT1 (Met1~Ala61), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

In man, metallothioneins are encoded by at least 10 to 12 genes separated into 2 groups designated MT-I and MT-II. Masters et al. (1994) noted that, unlike MT-I and MT-II, which are expressed in most organs, MT-III  expression appears to be restricted to the brain, and MT-IV is only expressed in certain stratified squamous epithelia. Class I metallothioneins contain 2 metal-binding domains: four divalent ions are chelated within cluster A of the alpha domain and are coordinated via cysteinyl thiolate bridges to 11 cysteine ligands. Cluster B, the corresponding region within the beta domain, can ligate three divalent ions to 9 cysteines.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: MT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR