Anti-GSTp Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB090HU01
MSPP-PAB090HU1EA 332 USD
MSPP-PAB090HU1
Anti-GSTp Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Glutathione S Transferase Pi (GSTp), derived from recombinant GSTp (Met1~Gln210), is reactive with Human/Mouse/Rat/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, the soluble GSTs are categorized into four main classes: alpha, mu, pi, and theta. The glutathione S-transferase pi gene (GSTP1) is a polymorphic gene encoding active, functionally different GSTP1 variant proteins that are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases. The predicted 209-amino acid protein shares 86% sequence identity with GSTP. However, GST-pi has a pI of 5.5, while that of GSTP is 6.9. Northern hybridization revealed that GST-pi is expressed as a 750-nucleotide mRNA in liver.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: GSTp
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR