Anti-SCT Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB075MU01
MSPP-PAB075MU1EA 342 USD
MSPP-PAB075MU1
Anti-SCT Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Secretin (SCT), derived from recombinant SCT (Pro26~Arg133), is reactive with Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Secretin is a hormone that controls the secretions into the duodenum, and also separately, water homeostasis throughout the body. Acting as an anti-Müllerian hormone it is produced in the S cells of the duodenum in the crypts of Lieberkühn. Its effect is to regulate the pH of the duodenal contents via the control of gastric acid secretion and buffering with bicarbonate. It is notable for being the first hormone to be identified. In humans, the secretin peptide is encoded by the SCT gene. It has recently been discovered to play a role in osmoregulation in the hypothalamus, pituitary, and kidney. Secretin is a linear peptide hormone, which is composed of 27 amino acids and has a molecular weight of 3055. A helix is formed in the amino acids between positions 5 and 13.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: SCT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR