Anti-PIGR Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB074MU03
MSPP-PAB074MU3EA 342 USD
MSPP-PAB074MU3
Anti-PIGR Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Polymeric Immunoglobulin Receptor (PIGR), derived from recombinant PIGR(Pro463~Ala563), is reactive with Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Polymeric immunoglobulin receptor is a Fc receptor which facilitates the secretion of IgA and IgM. The poly-Ig receptor is expressed on several glandular epithelia including those of liver and breast. It mediates transcellular transport of polymeric immunoglobulin molecules.,It is a member of the immunoglobulin superfamily (Hood et al., 1985). The receptor has 5 units with homology to the variable (V) units of immunoglobulins and a transmembrane region, which also has some homology to certain immunoglobulin variable regions.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PIGR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR