Anti-ITGb1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB042HU01
MSPP-PAB042HU1EA 332 USD
MSPP-PAB042HU1
Anti-ITGb1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Integrin Beta 1 (ITGb1), derived from recombinant ITGb1 (Asp140~Ile383), is reactive with Human/Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12), also known as ITGB1 or CD29, is a human gene. CD29 is an integrin unit associated with very late antigen receptors. Integrins are heterodimeric proteins made up of alpha and beta subunits. At least 18 alpha and 8 beta subunits have been described in mammals. Integrin family members are membrane receptors involved in cell adhesion and recognition in a variety of processes including embryogenesis, hemostasis, tissue repair, immune response and metatastatic diffusion of tumor cells. The protein encoded by this gene is a beta subunit. Six alternatively spliced variants have been found for this gene which encode five proteins with alternate carboxy termini.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: ITGb1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR