Anti-ENG Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA980RA01
MSPP-PAA980RA1EA 342 USD
MSPP-PAA980RA1
Anti-ENG Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Endoglin (ENG), derived from recombinant ENG(Glu26~Pro136), is reactive with Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Endoglin has a calculated monomeric molecular mass of 61 kDa but as a result of glycosylation, migrates at approximately 75 to 85 kDa under reducing conditions in SDS-PAGE. Endoglin, also known as CD105, is a Type I integral membrane glycoprotein with a large, disulfide-linked, extracellular region and a short, constitutively phosphorylated, cytoplasmic tail. Two splice variants of human endoglin, the S-endoglin and L-endoglin that differ in the length of their cytoplasmic tails have been identified. Endoglin is highly expressed on vascular endothelial cells, chondrocytes, and syncytiotrophoblasts of term placenta. It is also found on activated monocytes, bone marrow pro-erythroblasts, and leukemic cells of lymphoid and myeloid lineages.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: ENG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR