Anti-NB1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA943MU01
MSPP-PAA943MU1EA 324 USD
MSPP-PAA943MU1
Anti-NB1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Neutrophil Specific Antigen 1 (NB1), derived from recombinant NB1 (Ala193~Arg468), is reactive with Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

NB1, a glycosyl-phosphatidylinositol (GPI)-linked N-glycosylated cell surface glycoprotein, was first described in a case of neonatal alloimmune neutropenia. Independently, Kissel et al., (2001) cloned and characterized NB1, which they also referred to as CD177 and HNA2A. They obtained the cDNA after purification and microsequence analysis of NB1 protein from normal resting granulocytes. Kissel et al., (2001) noted amino acid differences between the PRV1 (Temerinac et al., 2000) and NB1 sequences at positions 3, 119, 323, and 379 and suggested that 2 different, highly homologous genes may exist. By genomic sequence analysis and PCR, Kissel et al., (2002) determined that the NB1 gene contains 9 exons.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: NB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR