Anti-Anti-INSR Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA895HU02
MSPP-PAA895HU2EA 316 USD
MSPP-PAA895HU2
Anti-Anti-INSR Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Insulin Receptor (INSR), derived from recombinant Anti-INSR(Arg1027~Met1364), is reactive with Human/Mouse/Rat/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

In molecular biology, the insulin receptor is a transmembrane receptor that is activated by insulin. It belongs to the large class of tyrosine kinase receptors. Two alpha subunits and two beta subunits make up the insulin receptor. The beta subunits pass through the cellular membrane and are linked by disulfide bonds. The alpha and beta subunits are encoded by a single gene (INSR). The insulin receptor has also recently been designated CD220 .The main activity of activation of the insulin receptor is inducing glucose uptake. For this reason 'insulin insensitivity', or a decrease in insulin receptor signaling, leads to diabetes mellitus type 2 - the cells are unable to take up glucose, and the result is hyperglycemia (an increase in circulating glucose), and all the sequelae which result from diabetes.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: Anti-INSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR