Anti-PTH Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA866RA01
MSPP-PAA866RA1EA 342 USD
MSPP-PAA866RA1
Anti-PTH Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Parathyroid Hormone (PTH), derived from recombinant PTH (Ala32~Gln115), is reactive with Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Parathyroid hormone is secreted by the parathyroid glands as a polypeptide containing 84 amino acids. It acts to increase the concentration of calcium (Ca²⁺) in the blood, whereas calcitonin (a hormone produced by the parafollicular cells (C cells) of the thyroid gland) acts to decrease calcium concentration. PTH acts to increase the concentration of calcium in the blood by acting upon parathyroid hormone receptor in three parts of the body: PTH half-life is approximately 4 minutes. It has a molecular mass of 9.4 kDa. PTH was one of the first hormones to be shown to use the G-protein, adenylyl cyclase second messenger system. Normal total plasma calcium level ranges from 8.5 to 10.2 mg/dl (2.12 mmol/L to 2.55 mmol/L).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PTH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR