Anti-GnRH Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA843GA01
MSPP-PAA843GA1EA 342 USD
MSPP-PAA843GA1
Anti-GnRH Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Gonadotropin Releasing Hormone (GnRH), derived from recombinant GnRH (Gln24~Ile92), is reactive with Chicken.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Gonadotropin-releasing hormone (GnRH),is a tropic peptide hormone responsible for the release of FSH and LH from the anterior pituitary. GnRH is synthesized and released from neurons within the hypothalamus.The vertebrate gonadotropin-releasing hormone (GnRH) is a decapeptide involved in regulating reproduction. FSH and LH are secreted by the basophilic cells of the anterior pituitary in response to the Gonadotropin-releasing hormone (GnRH) produced by the hypothalamus. The release of GnRH from the hypothalamus regulates the production of gonadotropins in the pituitary and these gonadotropins are responsible for gonadal development and growth in vertebrates. In addition to the hypothalamic GnRH (also called GnRH1 or GnRH-I), many vertebrate species have been found to express other GnRH forms.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: GnRH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR