Anti-Lpa Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA842HU01
MSPP-PAA842HU1EA 316 USD
MSPP-PAA842HU1
Anti-Lpa Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Lipoprotein, a (Lpa), derived from recombinant Lpa(Asp1719~Arg2038), is reactive with Human/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Lipoprotein(a) consists of an LDL-like particle and the specific apolipoprotein(a) , which is covalently bound to the apoB of the LDL like particle. Lp(a) plasma concentrations are highly heritable and mainly controlled by the apolipoprotein(a) gene located on chromosome 6q26-27. Apo(a) proteins vary in size due to a size polymorphism , which is caused by a variable number of so called kringle IV repeats in the LPA gene. This size variation at the gene level is expressed on the protein level as well, resulting in apo(a) proteins with 10 to > 50 kringle IV repeats . These variable apo(a) sizes are known as 'apo(a) isoforms'. There is a general inverse correlation between the size of the apo(a)isoform and the Lp(a) plasma concentration which is caused by a variable rate of degradation before the apo(a) protein has matured for Lp(a) assembly.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: Lpa
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR