Anti-NOS2 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA837RA01
MSPP-PAA837RA1EA 342 USD
MSPP-PAA837RA1
Anti-NOS2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Nitric Oxide Synthase 2, Inducible (NOS2), derived from recombinant NOS2 (Ala518~Arg694), is reactive with Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Nitric oxide synthase 2A (inducible, hepatocytes)is an enzyme encoded by the NOS2 gene. is produced by a group of enzymes called nitric oxide synthases (NOS). These enzymes catalyze the production of NO and L-citrulline from L-arginine, O2, and NADPH-derived electrons. Mammalian systems contain three well-characterized isoforms of the enzyme: neuronal NOS (nNOS, also called NOS1), inducible NOS (iNOS or NOS2), and endothelial NOS (eNOS or NOS3). The names reflect characteristics of the activity or the original tissues in which the enzymes were first described, but it is now known that each of these isoforms is expressed in a variety of tissues and cell types. The three main isoforms share structural similarities and have nearly identical catalytic mechanisms.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: NOS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR