Anti-HIF1a Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA798GA03
MSPP-PAA798GA3EA 342 USD
MSPP-PAA798GA3
Anti-HIF1a Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Hypoxia Inducible Factor 1 Alpha (HIF1a), derived from recombinant HIF1a(Glu35~Val155), is reactive with Chicken.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The HIF (hypoxia-inducible factor) transcription factor is a member of the basic-helix-loop-helix (bHLH) family of transcription factors and plays an important role in maintaining cellular oxygen homeostasis. The hypoxia-inducible transcription factor 1α (HIF-1α) is the regulated member of the transcription factor heterodimer HIF-1. HIF-1α has emerged as an important drug target in breast and prostate cancer, cardiovascular disease, and ischemia.,HIF-1alpha abundance (and its subsequent activity) is regulated transcriptionally in an NF-κB dependent manner. In addition, the coordinated activity of the prolyl hydroxylases (PHDs) maintain the appropriate balance of HIF-1α protein in the post-translation phase.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: HIF1a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR