Anti-CDKN2A Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA794HU01
MSPP-PAA794HU1EA 316 USD
MSPP-PAA794HU1
Anti-CDKN2A Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Cyclin Dependent Kinase Inhibitor 2A (CDKN2A), derived from recombinant CDKN2A (Ser8~Asp156), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

CDKN2A, is a tumor suppressor protein, which in humans is encoded by the CDKN2A gene.P16 plays an important role in regulating the cell cycle, and mutations in p16 increase the risk of developing a variety of cancers, notably melanoma. This gene generates several transcript variants which differ in their first exons. At least three alternatively-spliced variants encoding distinct proteins have been reported, two of which encode structurally related isoforms known to function as inhibitors of CDK4 kinase. The remaining transcript includes an alternate first exon located 20 kb upstream of the remainder of the gene; this transcript contains an alternate open reading frame (ARF) that specifies a protein which is structurally unrelated to the products of the other variants.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: CDKN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR