Anti-PDL1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA788RA01
MSPP-PAA788RA1EA 342 USD
MSPP-PAA788RA1
Anti-PDL1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Programmed Cell Death Protein 1 Ligand 1 (PDL1), derived from recombinant PDL1 (Ser34~Val241), is reactive with Rat/Human/Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

CD274 (Cluster of Differentiation 274) also known as PD-L1 (Programed Death Ligand-1) or B7-H1 is a protein,PD-L1 binds to its receptor, PD-1, found on activated T cells, B cells, and myeloid cells, to modulate activation or inhibition. The affinity between PD-L1 and PD-1, as defined by the dissociation constant Kd, is 770 nM. Interestingly, PD-L1 also has an appreciable affinity for the costimulatory molecule CD80 (B7-1), but not CD86 (B7-2)., CD80's affinity for PD-L1, 1.4 µM, is intermediate between its affinities for CD28 and CTLA-4 (4.0 µM and 400 nM, respectively). The related molecule PD-L2 has no such affinity for CD80 or CD86, but shares PD-1 as a receptor (with a stronger kDa of 140 nM).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PDL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat;Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR