Anti-CS Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA723GE01
MSPP-PAA723GE1EA 398 USD
MSPP-PAA723GE1
Anti-CS Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Chondroitin Sulfate (CS), derived from BSA conjugated CS, is reactive with General species.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Chondroitin sulfate (CS) is a major microenvironmental molecule in the CNS, and there have been few reports about its neuroprotective activity. One of several classes of sulfated glycosaminoglycans that is a major constituent in various connective tissues, especially in the ground substance of blood vessels, bone, and cartilage.,Chondroitin sulfate is a sulfated glycosaminoglycan (GAG) composed of a chain of alternating sugars (N-acetylgalactosamine and glucuronic acid). It is usually found attached to proteins as part of a proteoglycan. A chondroitin chain can have over 100 individual sugars, each of which can be sulfated in variable positions and quantities. Understanding the functions of such diversity in chondroitin sulfate and related glycosaminoglycans is a major goal of glycobiology. Chondroitin sulfate is an important structural component of cartilage and provides much of its resistance to compression .

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: CS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: General species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR