Anti-ATP1b3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA712MU01
MSPP-PAA712MU1EA 324 USD
MSPP-PAA712MU1
Anti-ATP1b3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Sodium/potassium Transporting ATPase Subunit Beta-3 (ATP1b3), derived from recombinant ATP1b3(Met61~Arg277), is reactive with Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Sodium/potassium-transporting ATPase subunit beta-3 belongs to the family of Na /K and H /K ATPases beta chain proteins, and to the subfamily of Na+/K+-ATPases. Na+/K+-ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: ATP1b3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR