Anti-COX 2 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA699HU01
MSPP-PAA699HU1EA 316 USD
MSPP-PAA699HU1
Anti-COX 2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Cyclooxygenase-2 (COX 2), derived from recombinant COX 2 (Phe187~Asn425), is reactive with Human/Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Cyclooxygenase (COX) enzymes catalyze the synthesis of prostaglandins (PGs) from arachidonic acid. There are two isoforms of COX, COX-1 and COX-2. While COX-1 is expressed constitutively and appears to be responsible for the production of prostaglandins (PGs) that control normal physiologic functions, expression of COX-2 is induced by various inflammatory and mitogenic stimuli such as cytokines, growth factors or tumor promoters. Numerous experimental studies suggest a relationship between COX-2 expression and carcinogenesis. For example, increased amounts of COX-2 have been observed in breast cancers that overexpress HER-2/neu. Furthermore, treatment with selective inhibitors of COX-2 reduced the formation of tongue, esophagal, intestinal, breast, skin, lung, and bladder tumors in experimental animals.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: COX 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR