Anti-TLR2 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA663MU01
MSPP-PAA663MU1EA 324 USD
MSPP-PAA663MU1
Anti-TLR2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Toll Like Receptor 2 (TLR2), derived from recombinant TLR2(Gln25~Pro250), is reactive with Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

TLR-2 plays a role in the immune system. TLR-2 is a membrane protein, a receptor, which is expressed on the surface of certain cells and recognizes foreign substances and passes on appropriate signals to the cells of the immune system. TLR-2 is a membrane receptor found at the surface of immune system cells that recognises many bacterial, fungal, viral and endogenous substances. Phagocytosis of bound materials takes place in endosome/phagosome and a cellular activation, so that the elements of the innate immune system take over such as macrophages, PMN and dendritic cells tasks of the nonspecific immune defense, B1a and form MZB first anti-bodies and uses in the process the specific anti-body formation.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: TLR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR