Anti-NTXI Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA639HU01
MSPP-PAA639HU1EA 314 USD
MSPP-PAA639HU1
Anti-NTXI Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Cross Linked N-Telopeptide Of Type I Collagen (NTXI), derived from recombinant NTXI(Gln162~Pro178), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Type I collagen is a heterotrimer consisting of two α1(I) chains and one α2(I) chain. NTx assays are sensitive and specific assays that can be used to indicate subtle changes in levels of bone resorption. NTx serum and urine assays provide a quantitative measurement of the cross-linked N-telopeptides of bone type I collagen (NTx). NTx is a specific biochemical indicator of bone resorption that is generated as a result of osteoclast activity on bone. The NTx molecule is specific to bone due to the unique amino acid sequence and orientation of the cross-linked alpha-2 (I) N-telopeptide. NTx provides a dynamic indicator of the current level of bone resorption, shows significantly earlier response to therapy (with 3 to 6 months) and, when used in conjunction with bone mineral density (BMD), gives a more complete picture of bone status than either technology alone.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: NTXI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR