Anti-PCNA Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA591MI01
MSPP-PAA591MI1EA 368 USD
MSPP-PAA591MI1
Anti-PCNA Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Proliferating Cell Nuclear Antigen (PCNA), derived from recombinant PCNA (Leu6~Ser261), is reactive with Human/Mouse/Rat/Rabbit/Simian/Dog/Pig/Bovine/Goat/Sheep/Horse/Chicken.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Proliferating Cell Nuclear Antigen a protein that acts as a processivity factor for DNA polymerase delta in eukaryotic cells. It achieves this processivity by encircling the DNA, thus creating a topological link to the genome. It is an example of a DNA clamp. The protein encoded by this gene is found in the nucleus and is a cofactor of DNA polymerase delta. The encoded protein acts as a homotrimer and helps increase the processivity of leading strand synthesis during DNA replication. In response to DNA damage, this protein is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Two transcript variants encoding the same protein have been found for this gene. Pseudogenes of this gene have been described on chromosome 4 and on the X chromosome.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Rabbit;Simian;Dog;Pig;Bovine;Goat;Sheep;Horse;Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR