Anti-CCND1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA585HU01
MSPP-PAA585HU1EA 316 USD
MSPP-PAA585HU1
Anti-CCND1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Cyclin D1 (CCND1), derived from recombinant CCND1 (Met1~Ile295), is reactive with Human/Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Cyclin D1 belongs to the family of cyclin proteins which function as the regulatory subunits of cyclin/cyclin dependent kinase (Cdk) holoenzymes that regulate entry into and progression through the cell cycle. Cyclin D1 expression is induced upon stimulation by growth factors (e.g.EGF, IGF-I/II), amino acids, hormones, and oncogenes such as Ras, Src, ErbB2, and SV40 T antigen. Cdk4 and Cdk6 can partner with Cyclin D1 in early to mid-G1 phase to phosphorylate and inactivate the cell-cycle inhibitory function of the retinoblastoma protein (pRB) in cooperation with Cyclin E/Cdk2. Cyclin D1 is also known to modulate local chromatin structure and transcription of genes involved in proliferation and differentiation through CDK-independent association with histone acetylases (e.g. CBP, P/CAF) and deacetylases.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: CCND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR