Anti-IL1b Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA563RA01
MSPP-PAA563RA1EA 266 USD
MSPP-PAA563RA1
Anti-IL1b Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Interleukin 1 Beta (IL1b), derived from recombinant IL1b (Val117~Ser268), is reactive with Human/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Interleukin-1 (IL-1) is one of the first cytokines ever described. Its initial discovery was as a factor that could induce fever, control lymphocytes, increase the number of bone marrow cells and cause degeneration of bone joints. At this time, IL-1 was known under several other names including endogenous pyrogen, lymphocyte activating factor, haemopoetin-1 and mononuclear cell factor, amongst others., It was around 1984 to 1985 when scientists confirmed that IL-1 was actually composed of two distinct proteins, now called IL-1α and IL-1β.These belong to a family of cytokines known as the interleukin-1 superfamily.Both IL-1α and IL-1β are produced by macrophages, monocytes and dendritic cells.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IL1b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR