Anti-EGF Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA560PO01
MSPP-PAA560PO1EA 378 USD
MSPP-PAA560PO1
Anti-EGF Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Epidermal Growth Factor (EGF), derived from recombinant EGF (Ala45~His275), is reactive with Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Epidermal growth factor (EGF) was discovered in crude preparations of nerve growth factor prepared from mouse submaxillary glands as an activity that induced early eyelid opening, incisor eruption, hair growth inhibition, and stunting of growth when injected into newborn mice.EGF is a member of a family of growth factors that bind to the same 170 kDa receptor, including TGF-, vaccinia growth factor and amphiregulin. EGF is initially synthesized as a large (130 kDa) precursor molecule in which the mature, soluble EGF sequence (6 kDa) is located. The precursor, which functions as a source for soluble EGF, may also have a role in mediation of intercellular communication between cells displaying pro-EGF on their surfaces and cells with EGF receptors.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR