Anti-MMP9 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA553HU01
MSPP-PAA553HU1EA 340 USD
MSPP-PAA553HU1
Anti-MMP9 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Matrix Metalloproteinase 9 (MMP9), derived from recombinant MMP9 (Gly213~Ala399), is reactive with Human/Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

MMP-9 (also known as gelatinase B) is secreted as a 92 kDa zymogen. Cleavage of pro-MMP-9 at or near residue 87 results in the active enzyme with a mass of approximately 82 kDa. MMP-9 has three fibronectin type II domains, a hemopexin-like domain and a proline-rich type V collagen-homologous domain., Pro-MMP-9 can be activated by MMP-3 or by certain bacterial proteinases. MMP-9 is inhibited byα2-macroglobulin or by TIMP-1, which binds to pro-MMP-9 as well as to active MMP-9.Pro-MMP-9 is secreted by monocytes, macrophages, neutrophils, keratinocytes, fibroblasts, osteoclasts, chondrocytes, skeletal muscle satellite cells, endothelial cells, and various tumor cells. Pro-MMP-9 expression is upregulated by TGF-β1, IL-1β, TGF-α, PDGF-AB.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR