Anti-INS Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA448HU01
MSPP-PAA448HU1EA 240 USD
MSPP-PAA448HU1
Anti-INS Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Insulin (INS), derived from recombinant INS(Phe25~Asn110), is reactive with Human/Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Insulin is a polypeptide hormone originating in the beta cells of the pancreas and serving as a principal regulator for the storage and production of carbohydrates. Its secretion is normally stimulated by increases in the amount of glucose in circulation. This leads to higher insulin levels and more rapid tissueassimilation of glucose followed by a decline in the insulin level as the glucose level subsides. In a number of conditions, notably insulinoma and diabetes, this relationship is impaired. Insulin tends to circulate at inappropriately high levels in patients with insulin-secreting pancreatic tumors; such tumors can thus be a cause of hypoglycemia. Accordingly, insulin immunoassays used sometimes in connection with provocative doses of tolbutamide or calcium play an essential role in the identification (and localization) of insulinomas.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: INS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR