Anti-PKCe Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA439HU01
MSPP-PAA439HU1EA 316 USD
MSPP-PAA439HU1
Anti-PKCe Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Protein Kinase C Epsilon (PKCe), derived from recombinant PKCe(Leu403~Asp672), is reactive with Human/Mouse/Rat/Pig.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. Protein kinase C epsilon type kinase has been shown to be involved in many different cellular functions, such as neuron channel activation, apoptosis, cardioprotection from ischemia, heat shock response, as well as insulin exocytosis. Knockout studies in mice suggest that this kinase is important for lipopolysaccharide (LPS)-mediated signaling in activated macrophages and may also play a role in controlling anxiety-like behavior.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PKCe
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR