Anti-EMILIN1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA415RA01
MSPP-PAA415RA1EA 342 USD
MSPP-PAA415RA1
Anti-EMILIN1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Elastin Microfibril Interface Located Protein 1 (EMILIN1), derived from recombinant EMILIN1 (Leu684~Gln899), is reactive with Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

EMILIN (elastin microfibril interface located protein) is an extracellular matrix glycoprotein abundantly expressed in elastin-rich tissues such as blood vessels, skin, heart, and lung., The extracellular matrix protein EMILIN1 (elastin microfibril interface located protein 1) is implicated in maintaining blood pressure homeostasis via the N-terminal elastin microfibril interface domain and in trophoblast invasion of the uterine wall via the globular C1q (gC1q) domain. Elastin microfibril interface-located proteins (EMILINs) constitute a family of extracellular matrix (ECM) glycoproteins characterized by the presence of an EMI domain at the N terminus and a gC1q domain at the C terminus.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: EMILIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR