Anti-C5a Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA388MU01
MSPP-PAA388MU1EA 288 USD
MSPP-PAA388MU1
Anti-C5a Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Complement Component 5a (C5a), derived from recombinant C5a(Asn679~Arg755), is reactive with Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

C5a is a protein fragment released from complement component C5. In humans, the polypeptide contains 74 amino acids. NMR spectroscopy proved that the molecule is composed of four helices and loops connecting the helices. On the N terminus a short 1.5 turn helix is also present. The longest helix -IV- develops three disulfide bonds with helix II and III. C5a is rapidly metabolised by a serum enzyme, carboxypeptidase B to a 73 amino acid form, C5a des-Arg.

C5a binds to a receptor protein on the surface of target cells, C5aR or CD88. This is a member of the G-protein-coupled receptor superfamily of proteins, predicted to have seven transmembrane helical domains of largely hydrophobic amino acid residues, forming three intra- and three extra-cellular loops, with an extracellular N-terminus and an intracellular C-terminus.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: C5a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR