Anti-RBP2 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA377HU01
MSPP-PAA377HU1EA 316 USD
MSPP-PAA377HU1
Anti-RBP2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Retinol Binding Protein 2, Cellular (RBP2), derived from recombinant RBP2 (Met1~Lys134), is reactive with Human/Mouse/Rat/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Cellular retinol-binding protein-2 is an abundant 134-residue protein present in the small intestinal epithelium. It is thought to participate in the uptake and/or intracellular metabolism of vitamin A. Demmer et al. (1987) isolated and sequenced the rat Crbp2 gene. Its 4 exons span 0.65 Kb and are interrupted by 3 introns within an aggregate length of 19.5 Kb. Southern blot hybridization analysis indicated that the gene is highly conserved in rats, mice, and humans. Crbp2 belongs to a protein family that contains liver fatty acid-binding protein. Demmer et al. (1987) mapped the gene for Crbp2 in mice using recombinant inbred strains and restriction fragment length polymorphisms. They were found to be located on chromosome 9 within 5.3 cm of the phosphoglucomutase-3 locus and closely linked to Crbp, which is highly homologous.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: RBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR