Anti-RBP3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA367RA02
MSPP-PAA367RA2EA 342 USD
MSPP-PAA367RA2
Anti-RBP3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Retinol Binding Protein 3, Interstitial (RBP3), derived from recombinant RBP3(Ile1027~Gln1229), is reactive with Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Retinol-binding protein 3, interstitial (RBP3), also known as IRBP is a protein identified in most eutherians except tenrecs and armadillos. The interphotoreceptor retinol-binding protein is a large glycoprotein known to bind retinoids and found primarily in the interphotoreceptor matrix of the retina between the retinal pigment epithelium and the photoreceptor cells. It is thought to transport retinoids between the retinal pigment epithelium and the photoreceptors, a critical role in the visual process. The human IRBP gene is approximately 9.5 kbp in length and consists of four exons separated by three introns. The introns are 1.6-1.9 kbp long. The gene is transcribed by photoreceptor and retinoblastoma cells into an approximately 4.3-kilobase mRNA that is translated and processed into a glycosylated protein of 135,000 Da.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: RBP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR