Anti-PKIb Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA359HU01
MSPP-PAA359HU1EA 314 USD
MSPP-PAA359HU1
Anti-PKIb Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Protein Kinase Inhibitor Beta (PKIb), derived from recombinant PKIb (Met8~Glu76), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

cAMP-dependent protein kinase inhibitor beta is a member of the cAMP-dependent protein kinase inhibitor family. Studies of a similar protein in rat suggest that this protein may interact with the catalytic subunit of cAMP-dependent protein kinase and act as a competitive inhibitor. At least three alternatively spliced transcript variants encoding the same protein have been reported. The deduced 70-amino acid protein has a calculated molecular mass of about 7.4 kD. The purified protein has an apparent molecular mass of 10.5 by SDS-PAGE. Using kinetic analysis for inhibitor activity, Van Patten et al., (1991) determined that purified PKIB is a competitive inhibitor of cAMP-dependent protein kinase.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PKIb
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR