Anti-APOC1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA252MU01
MSPP-PAA252MU1EA 324 USD
MSPP-PAA252MU1
Anti-APOC1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Apolipoprotein C1 (APOC1), derived from recombinant APOC1 (Gly32~Ser88), is reactive with Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Apolipoprotein C1 is a lipoprotein with a length of 57 amino acids normally found in plasma and responsible for the activation of esterified lechitin cholesterol with an important role in the exchange of esterified cholesterol between lipoproteins and in removal of cholesterol from tissues., Its main function is inhibition of CETP, probably by altering the electric charge of HDL molecules. during fasting (like other apolipoprotein C) is found primarily within HDL, meanwhile after a meal it is found within the surface of other lipoproteins. in contrast when proteins rich in triglycerides like chylomicrons and VLDL are broken down this apoprotein is transferred again to the HDL.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: APOC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR