Anti-AMH Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA228FI08
MSPP-PAA228FI8EA 450 USD
MSPP-PAA228FI8
Anti-AMH Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Anti-Mullerian Hormone (AMH), derived from recombinant AMH (Thr118~Arg134), is reactive with Zebrafish.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Anti-Müllerian hormone (AMH) is a dimeric glycoprotein that inhibits the development of the Müllerian ducts in a male embryo. It is named after Johannes Peter Müller. AMH is a protein hormone structurally related to inhibin and activin, and a member of the transforming growth factor-β (TGF-β) family. It is present in fish, reptiles, birds, marsupials, and placental mammals. In mammals AMH is secreted by Sertoli cells of the testes during embryogenesis of the fetal male and prevents the development of the mullerian ducts into the uterus and other mullerian structures. The effect is ipsilateral, that is each testis suppresses Müllerian development only on its own side. In humans this action takes place by eight weeks gestation. In female embryogenesis the absence of AMH allows for the development of upper vagina, uterus and cervix, and oviducts.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: AMH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR