Anti-ANP Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA225RA02
MSPP-PAA225RA2EA 342 USD
MSPP-PAA225RA2
Anti-ANP Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Atrial Natriuretic Peptide (ANP), derived from recombinant ANP, is reactive with Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

ANP is a cardiac hormone involved in the regulation of blood pressure and fluid balance. It is primarily produced by the heart's atria and is released in response to atrial stretch. ANP functions by promoting vasodilation, increasing renal sodium excretion, and reducing blood volume, which collectively lower blood pressure. It acts through specific receptors that stimulate cyclic GMP production, leading to its physiological effects. ANP is an important biomarker for heart failure and other cardiovascular conditions, given its role in modulating cardiovascular and renal function.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: ANP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR