Anti-ALT Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA207HU02
MSPP-PAA207HU2EA 316 USD
MSPP-PAA207HU2
Anti-ALT Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Alanine Aminotransferase (ALT), derived from recombinant ALT(Ile147~Phe263), is reactive with Human/Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Alanine transaminase or ALT is a transaminase enzyme. It is also called serum glutamic pyruvic transaminase (SGPT) or alanine aminotransferase (ALAT). ALT is found in serum and in various bodily tissues, but is most commonly associated with the liver. It catalyzes the transfer of an amino group from alanine to a-ketoglutarate, the products of this reversible transamination reaction being pyruvate and glutamate. It is commonly measured clinically as a part of a diagnostic liver function test, to determine liver health. GPT1 transcript is moderately expressed in kidney, liver, heart, and fat. Another GPT1 transcript shows a slightly different expression pattern, with a 3.9-kb transcript expressed at high levels in muscle, fat, kidney, and brain and at lower levels in liver and breast.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: ALT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR