Anti-CEA Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA150HU04
MSPP-PAA150HU4EA 210 USD
MSPP-PAA150HU4
Anti-CEA Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Carcinoembryonic Antigen (CEA), derived from recombinant CEA(Ser448~Gly572), is reactive with Human/Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Carcinoembryonic antigen (CEA) is a cell-surface glycoprotein with a molecular weight of 180-200kD, that occurs in high levels in colon epithelial cells during embryonic development. Levels of CEA are significantly lower in colon tissue of adults, but can become elevated when inflammation or tumours arise in any endodermal tissue, including the gastrointestinal tract, respiratory tract, pancreas and breast. An overexpression of CEA protein has been detected in a variety of adenocarcinomas, including gastric, pancreatic, small intestine, colon, rectal, ovarian, breast, cervical and non-small-cell lung cancers. CEA is also expressed by epithelial cells in several non-malignant disorders, including diverticulitis, pancreatitis, inflammatory bowel disease, cirrhosis, hepatitis, bronchitis and renal failure and also in heavy smokers.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: CEA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR