Anti-COL4a1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA149MU01
MSPP-PAA149MU1EA 324 USD
MSPP-PAA149MU1
Anti-COL4a1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Collagen Type IV Alpha 1 (COL4a1), derived from recombinant COL4a1 (Pro1416~Thr1669), is reactive with Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Collagen, type IV, alpha 1 is a major type IV alpha collagen chain of basement membranes. Like the other members of the type IV collagen gene family, this gene is organized in a head-to-head conformation with another type IV collagen gene so that each gene pair shares a common promoter. Types I, II, and III collagen, the so-called interstitial collagens, are in many ways distinct from basement membrane collagen. Type IV collagen does not form ordered fibrillar structures; rather, a meshwork is formed by 4 molecules held together at the ends. Both disulfide and typical lysyl-derived collagen crosslinks are involved. The collagen IV molecule is a heterotrimer of 2 alpha-1 chains and 1 alpha-2 chain. The alpha-2 chain is encoded at a distinct locus (COL4A2).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: COL4a1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR