Anti-VEGFA Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA143HU01
MSPP-PAA143HU1EA 280 USD
MSPP-PAA143HU1
Anti-VEGFA Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Vascular Endothelial Growth Factor A (VEGFA), derived from recombinant VEGFA(Pro28~Arg147), is reactive with Human/Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

VEGF-A is a glycosylated mitogen that specifically acts on endothelial cells and has various effects, including mediating increased vascular permeability, inducing angiogenesis, vasculogenesis and endothelial cell growth, promoting cell migration, and inhibiting apoptosis. Alternatively spliced transcript variants, encoding either freely secreted or cell-associated isoforms, have been characterized. VEGF is a chemical signal produced by cells that stimulates the growth of new blood vessels. The most important member is VEGF-A. Other members are Placenta growth factor (PlGF), VEGF-B, VEGF-C and VEGF-D. The latter ones were discovered later than VEGF-A, and, before their discovery, VEGF-A was called just VEGF.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: VEGFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR