Anti-MMP7 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA102HU01
MSPP-PAA102HU1EA 226 USD
MSPP-PAA102HU1
Anti-MMP7 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Matrix Metalloproteinase 7 (MMP7), derived from recombinant MMP7(Met1~Lys267), is reactive with Human/Mouse/Pig.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

MMP7, also known as Matrilysin, is the smallest member of the MMP-family, it is a zinc-dependent metalloproteinase and is overexpressed in colon cancer and many other human cancers. MMP7 is a secreted matrix metalloproteinase, which contributes to angiogenesis by breaking down basement membranes. Matrix Metalloproteinases (MMPs) belong to a family of secreted or membrane-associated zinc endopeptidases capable of digesting extracellular matrix components. MMP7 is responsible for hydrolyzing proteoglycans and ECM glycoproteins. It is expressed in normal cells, specifically glandular epithelial cells. MMP7 is thought to play an important role in the early steps of tumor progression. Expression of MMP2, MMP7 and MMP11 is greater in pancreatic carcinoma than in normal pancreas.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR