Anti-LIF Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA085HU01
MSPP-PAA085HU1EA 224 USD
MSPP-PAA085HU1
Anti-LIF Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Leukemia Inhibitory Factor (LIF), derived from recombinant LIF (Lys2~Ala201), is reactive with Human/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Leukemia inhibitory factor, an interleukin 6 class cytokine, is a protein in cells that affects cell growth and development. LIF derives its name from its ability to induce the terminal differentiation of myeloid leukaemic cells. Other properties attributed to the cytokine include: the growth promotion and cell differentiation of different types of target cells, influence on bone metabolism, cachexia, neural development, embryogenesis and inflammation. LIF is normally expressed in the trophectoderm of the developing embryo, with its receptor LIFR expressed throughout the inner cell mass. As embryonic stem cells are derived from the inner cell mass at the blastocyst stage, removing them from the inner cell mass also removes their source of LIF.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: LIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR