Anti-AngII Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA005HU01
MSPP-PAA05HU01EA 316 USD
MSPP-PAA05HU01
Anti-AngII Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Angiotensin II (AngII), derived from OVA conjugated AngII, is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Angiotensin I is converted to angiotensin II through removal of two terminal residues by the enzyme Angiotensin-converting enzyme (ACE, or kinase), which is found predominantly in the capillaries of the lung. ACE is actually found all over the body, but has its highest density in the lung due to the high density of capillary beds there. Angiotensin II acts as an endocrine, autocrine/paracrine, and intracrine hormone.ACE is a target for inactivation by ACE inhibitor drugs, which decrease the rate of angiotensin II production.,Angiotensin II increases blood pressure by stimulating the Gq protein in vascular smooth muscle cells . ACE inhibitor drugs are major drugs against hypertension.Other cleavage products of ACE, 7 or 9 amino acids long, are also known, they have differential affinity for angiotensin receptors, although their exact role is still unclear.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: AngII
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR