Anti-IL11 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA057MU01
MSPP-PAA057MU1EA 324 USD
MSPP-PAA057MU1
Anti-IL11 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Interleukin 11 (IL11), derived from recombinant IL11 (Pro25~Leu199), is reactive with Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Interleukins (IL) are a group of cytokines that play an important role in the immune system. They modulate inflammation and immunity by regulating growth, mobility and differentiation of lymphoid and other cells. Interleukin-11 (IL-11) is a pleiotropic cytokine that stimulates megakaryocytopoiesis, resulting in increased production of platelets, as well as activating osteoclasts, inhibiting epithelial cell proliferation and apoptosis, and inhibiting macrophage mediator production. These functions may be particularly important in mediating the hematopoietic, osseous and mucosal protective effects of IL-11. The cytokine also possesses anti-inflammatory activity, and has been proposed as a therapeutic agent in the treatment of chronic inflammatory diseases, such as Crohn's disease and rheumatoid arthritis.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR