Anti-LN Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA037MU01
MSPP-PAA037MU1EA 324 USD
MSPP-PAA037MU1
Anti-LN Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Laminin (LN), derived from native LN, is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Fibronectin (Fn) is a large modular glycoprotein that is found as a polymeric fibrillar network in the extracellular matirx (ECM) and as soluble disulfide-linked dimeric protomers in plasma and other body fluids. The protein subunit is made up of three types of homologous structural repeats termed Fn type I, type II, and type III repeats. Multiple isoforms of the protein formed by alternative splicing at numerous sites, resulting in insertions of extra type III domains (EDA and ECB) or parts of the variable type III connecting segment (V/IIICS), have been identified. Fibronectin is a ligand for fibrin, heparin, chondroitin sulfate, collagen/gelatin, and many integrin receptors. It is involved in multiple cellular processes including cell adhesion/migration, blood clotting, morphogenesis, tissue repair, and cell signaling.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: LN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR