Anti-PAPPA Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAA800MU01
MSPP-PA800MU01EA 324 USD
MSPP-PA800MU01
Anti-PAPPA Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Pregnancy Associated Plasma Protein A(PAPPA), derived from recombinant PAPPA(Phe999~Asn1272), is reactive with Mouse/Human/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

PAPP-A is a large heterottetrameric glycoprotein of approximately 500 kDa, which was first discovered in serum from pregnant women. The hetreotetramer consists of two PAPP-A subunits and two proMBP subunits. Low maternal serum levels of PAPP-A in first trimester biochemical screening is used as a marker of Down’s syndrome. a secreted metalloproteinase which cleaves insulin-like growth factor binding proteins (IGFBPs). It is thought to be involved in local proliferative processes such as wound healing and bone remodeling. Low plasma level of this protein has been suggested as a biochemical marker for pregnancies with aneuploid fetuses (fetuses with an abnormal number of chromosomes). For example, low PAPPA may be seen in prenatal screening for Down syndrome.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PAPPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR