Osmolality Standards, Wescor®
Supplier: ELITECHGROUP INC. MS
OA-010 OA-029 OA-100
MSPP-OA-010EA 173.85 USD
MSPP-OA-010 MSPP-OA-029 MSPP-OA-100
Osmolality Standards, Wescor®
Standards Osmolality Standards
Wescor ampule osmolality standards provide uncompromising accuracy to satisfy the most stringent laboratory quality assurance regimens. Calibration integrity is assured because ampules provide fresh solution with each use
  • Ampules provide fresh solution with each use
  • Ampule is fitted with a safety breaker sleeve
Delivery information: Each ampule contains 0.4 mL of solution.